Merck
  • Regulation of localization and function of the transcriptional co-activator YAP by angiomotin.
Regulation of localization and function of the transcriptional co-activator YAP by angiomotin.
eLife (2017-05-04)
Susana Moleirinho, Sany Hoxha, Vinay Mandati, Graziella Curtale, Scott Troutman, Ursula Ehmer, Joseph L Kissil
ABSTRACT
The Hippo-YAP pathway is a central regulator of cell contact inhibition, proliferation and death. There are conflicting reports regarding the role of Angiomotin (Amot) in regulating this pathway. While some studies suggest a YAP-inhibitory function other studies indicate Amot is required for YAP activity. Here, we describe an Amot-dependent complex comprised of Amot, YAP and Merlin. The phosphorylation of Amot at Serine 176 shifts localization of this complex to the plasma membrane, where it associates with the tight-junction proteins Pals1/PATJ and E-cadherin. Conversely, hypophosphorylated Amot shifts localization of the complex to the nucleus, where it facilitates the association of YAP and TEAD, induces transcriptional activation of YAP target genes and promotes YAP-dependent cell proliferation. We propose that phosphorylation of AmotS176 is a critical post-translational modification that suppresses YAP's ability to promote cell proliferation and tumorigenesis by altering the subcellular localization of an essential YAP co-factor.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Rac1/Cdc42 Activation Assay Kit, The Rac1/Cdc42 Activation Assay provides an effective method for detecting Rac & Cdc42 activity in cell lysates.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)