Merck
  • Cell-Type-Specific Translation Profiling Reveals a Novel Strategy for Treating Fragile X Syndrome.
Cell-Type-Specific Translation Profiling Reveals a Novel Strategy for Treating Fragile X Syndrome.
Neuron (2017-08-05)
Sophie R Thomson, Sang S Seo, Stephanie A Barnes, Susana R Louros, Melania Muscas, Owen Dando, Caoimhe Kirby, David J A Wyllie, Giles E Hardingham, Peter C Kind, Emily K Osterweil
ABSTRACT
Excessive mRNA translation downstream of group I metabotropic glutamate receptors (mGlu
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
(S)-3,5-Dihydroxyphenylglycine hydrate, ≥98% (HPLC), powder