Merck
Supraphysiological levels of GDF11 induce striated muscle atrophy.
EMBO molecular medicine (2017-03-09)
David W Hammers, Melissa Merscham-Banda, Jennifer Ying Hsiao, Stefan Engst, James J Hartman, H Lee Sweeney
ABSTRACT
Growth and differentiation factor (GDF) 11 is a member of the transforming growth factor β superfamily recently identified as a potential therapeutic for age-related cardiac and skeletal muscle decrements, despite high homology to myostatin (Mstn), a potent negative regulator of muscle mass. Though several reports have refuted these data, the