Merck
  • Cytoplasmic Relocalization of TAR DNA-Binding Protein 43 Is Not Sufficient to Reproduce Cellular Pathologies Associated with ALS
Cytoplasmic Relocalization of TAR DNA-Binding Protein 43 Is Not Sufficient to Reproduce Cellular Pathologies Associated with ALS
Frontiers in molecular neuroscience (2017-03-14)
Heike J Wobst, Steven S Wesolowski, Jayashree Chadchankar, Louise Delsing, Steven Jacobsen, Jayanta Mukherjee, Tarek Z Deeb, John Dunlop, Nicholas J Brandon, Stephen J Moss
ABSTRACT
Mutations in the gene