Merck
  • Nonalcoholic Steatohepatitis Modulates Membrane Protein Retrieval and Insertion Processes.
Nonalcoholic Steatohepatitis Modulates Membrane Protein Retrieval and Insertion Processes.
Drug metabolism and disposition: the biological fate of chemicals (2016-09-09)
A L Dzierlenga, J D Clarke, N J Cherrington
ABSTRACT
Interindividual variability in drug response in nonalcoholic steatohepatitis (NASH) can be mediated by altered regulation of drug metabolizing enzymes and transporters. Among these is the mislocalization of multidrug resistance-associated protein (MRP2)/Mrp2 away from the canalicular membrane, which results in decreased transport of MRP2/Mrp2 substrates. The exact mechanism of this mislocalization is unknown, although increased activation of membrane retrieval processes may be one possibility. The current study measures the activation status of various mediators implicated in the active membrane retrieval or insertion of membrane proteins to identify which processes may be important in rodent methionine and choline deficient diet-induced NASH. The mediators currently known to be associated with transporter mislocalization are stimulated by oxidative stressors and choleretic stimuli, which play a role in the pathogenesis of NASH. The activation of protein kinases PKA, PKCα, PKCδ, and PKCε and substrates radixin, myristoylated alanine-rich C-kinase substrate, and Rab11 were measured by comparing the expression, phosphorylation, and membrane translocation between control and NASH. Many of the mediators exhibited altered activation in NASH rats. Consistent with membrane retrieval of Mrp2, NASH rats exhibited a decreased phosphorylation of radixin and increased membrane localization of PKCδ and PKCε, thought to be mediators of radixin dephosphorylation. Altered activation of PKCδ, PKA, and PKCα may impair the Rab11-mediated active insertion of Mrp2. Overall, these data suggest alterations in membrane retrieval and insertion processes that may contribute to altered localization of membrane proteins in NASH.