Merck
  • Specificity in killing pathogens is mediated by distinct repertoires of human neutrophil peptides.
Specificity in killing pathogens is mediated by distinct repertoires of human neutrophil peptides.
Journal of innate immunity (2010-09-08)
Andreas Cederlund, Birgitta Agerberth, Peter Bergman
ABSTRACT
Neutrophil-derived antimicrobial peptides and proteins (AMPs) play an important role in the defense against microbes. Absence of defense is illustrated by neutropenic patients with frequent bacterial and fungal infections. However, the specificity of the antimicrobial effects has not been adequately described. We set out to determine the specific antimicrobial pattern of polypeptides in neutrophils (polymorphonuclear leukocytes, PMNs) against 4 potential human pathogens: Moraxella catarrhalis, Staphylococcus aureus, Haemophilus influenzae and Candida albicans. Protein extracts of human PMNs were separated using high-performance liquid chromatography and fractions were assayed for antimicrobial activity. Fractions displaying antimicrobial activity were separated on SDS-PAGE and characterized using MALDI-MS. Depletion experiments were utilized to determine the contribution of each AMP to the antimicrobial effect. Among the identified AMPs, α-defensins 1-3, azurocidin, LL-37, lysozyme, calprotectin and lactotransferrin were studied in detail. We found a divergent pattern of killing, that is, certain peptides and proteins exhibited selective activity against specific pathogens, while others displayed a broader antimicrobial activity. α-Defensins, LL-37 and calprotectin were active against all species, while lactotransferrin exclusively inhibited growth of S. aureus. Conversely, azurocidin was active against all species except S. aureus. Our observations may shed light on bacterial resistance to AMPs and on the elimination of specific bacterial communities on mucosal surfaces.