Merck
Deciphering caveolar functions by
eLife (2017-12-06)
Eric Seemann, Minxuan Sun, Sarah Krueger, Jessica Tröger, Wenya Hou, Natja Haag, Susann Schüler, Martin Westermann, Christian A Huebner, Bernd Romeike, Michael M Kessels, Britta Qualmann
ABSTRACT
Several human diseases are associated with a lack of caveolae. Yet, the functions of caveolae and the molecular mechanisms critical for shaping them still are debated. We show that muscle cells of
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid