Merck
  • The role of proto-oncogene GLI1 in pituitary adenoma formation and cell survival regulation.
The role of proto-oncogene GLI1 in pituitary adenoma formation and cell survival regulation.
Endocrine-related cancer (2015-07-30)
Katharina Lampichler, Patricio Ferrer, Greisa Vila, Mirjam I Lutz, Florian Wolf, Engelbert Knosp, Ludwig Wagner, Anton Luger, Sabina Baumgartner-Parzer
ABSTRACT
The Hedgehog (Hh) pathway is an important regulator of early tissue patterning and stem cell propagation. It was found to be aberrantly activated in numerous types of human cancer and might be relevant in cancer stem cells. The identification of adult stem cells in the pituitary raised the question if tumor-initiating cells and Hh signaling are involved in pituitary adenoma formation. The present study aimed at the evaluation of Hh signaling in relation to stem cell and cell cycle markers in 30 human pituitary adenomas and in cultured murine adenoma cells. Therefore, expression levels of components of the Hh pathway, stem cell marker SOX2, cell cycle regulator tumor-protein 53 (TP53), proliferation marker Ki67 (MKI67) and superoxide dismutase 1 (SOD1) were evaluated in 30 human pituitary adenomas in comparison to control tissue. Modulation of cell function and target gene expression by the inhibition and activation of the Hh pathway were studied in murine adenoma cells. We show that transcription factor glioma-associated oncogene 1 (GLI1) is overexpressed in 87% of all pituitary adenomas. The expression of GLI1 significantly correlated with that of SOX2, TP53, MKI67 and SOD1. Inhibition of GLI1 resulted in the downregulation of the above genes and severe cell death in mouse adenoma cells. On the other hand, activation of the Hh pathway increased cell viability and target gene expression. In conclusion, our findings point toward an alternative, ligand-independent Hh pathway activation with GLI1 playing a major role in the cell survival of pituitary adenoma cells.
Sigma-Aldrich
GANT61, ≥98% (HPLC)
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks