Merck
  • Targeting the cross-talk between Urokinase receptor and Formyl peptide receptor type 1 to prevent invasion and trans-endothelial migration of melanoma cells.
Targeting the cross-talk between Urokinase receptor and Formyl peptide receptor type 1 to prevent invasion and trans-endothelial migration of melanoma cells.
Journal of experimental & clinical cancer research : CR (2017-12-09)
Concetta Ragone, Michele Minopoli, Vincenzo Ingangi, Giovanni Botti, Federica Fratangelo, Antonello Pessi, Maria Patrizia Stoppelli, Paolo Antonio Ascierto, Gennaro Ciliberto, Maria Letizia Motti, Maria Vincenza Carriero
ABSTRACT
Accumulating evidence demonstrates that the Urokinase Receptor (uPAR) regulates tumor cell migration through its assembly in composite regulatory units with transmembrane receptors, and uPAR Expression levels of uPAR and FPR1 were assessed by immunocytochemistry, Western Blot and qRT-PCR. Cell migration was investigated by Boyden chamber and wound-healing assays. Migration and invasion kinetics, trans-endothelial migration and proliferation of melanoma cells were monitored in real time using the xCELLigence technology. The agonist-triggered FPR1 internalization was visualized by confocal microscope. Cell adhesion to endothelium was determined by fluorometer measurement of cell-associated fluorescence or identified on multiple z-series by laser confocal microscopy. The 3D-organotypic models were set up by seeding melanoma cells onto collagen I matrices embedded dermal fibroblasts. Data were analyzed by one-way ANOVA and post-hoc Dunnett t-test for multiple comparisons. We found that the co-expression of uPAR and FPR1 confers to A375 and M14 melanoma cells a clear-cut capability to move towards chemotactic gradients, to cross extracellular matrix and endothelial monolayers. FPR1 activity is required, as cell migration and invasion were abrogated by receptor desensitization. Finally, melanoma cell ability to move toward chemotactic gradients, invade matrigel or fibroblast-embedded collagen matrices and cross endothelial monolayers are prevented by anti-uPAR Collectively, our findings identify uPAR and FPR1 as relevant effectors of melanoma cell invasiveness and suggest that inhibitors of the uPAR
Sigma-Aldrich
Cell dissociation sieve - tissue grinder kit, tissue grinder homogenizer kit, stainless steel, autoclavable, 1/kit
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting human PLAUR
Sigma-Aldrich
Screens for CD-1, size 40 mesh