Merck
  • Boosting the efficiency of inverted quantum dot light-emitting diodes by balancing charge densities and suppressing exciton quenching through band alignment.
Boosting the efficiency of inverted quantum dot light-emitting diodes by balancing charge densities and suppressing exciton quenching through band alignment.
Nanoscale (2017-12-14)
Jiangyong Pan, Changting Wei, Lixi Wang, Jinyong Zhuang, Qianqian Huang, Wenming Su, Zheng Cui, Arokia Nathan, Wei Lei, Jing Chen
ABSTRACT
We report an inverted and multilayer quantum dot light emitting diode (QLED) which boosts high efficiency by tuning the energy band alignment between charge transport and light emitting layers. The electron transport layer (ETL) was ZnO nanoparticles (NPs) with an optimized doping concentration of cesium azide (CsN
Sigma-Aldrich
Dipyrazino[2,3-f:2′,3′-h]quinoxaline-2,3,6,7,10,11-hexacarbonitrile, 95% (HPLC)