Merck
  • PCAF-mediated acetylation of Lin28B increases let-7 biogenesis in lung adenocarcinoma H1299 cells.
PCAF-mediated acetylation of Lin28B increases let-7 biogenesis in lung adenocarcinoma H1299 cells.
BMC cancer (2018-01-06)
Ting-Ting Qu, Fei Chen, Jing Wang, Yan-Jun Zhang, Mo-Bin Cheng, Wen-Zheng Sun, Yu-Fei Shen, Ye Zhang
ABSTRACT
Lin28B and its paralog Lin28A are small RNA binding proteins that have similar inhibitory effects, although they target separate steps in the maturation of let-7 miRNAs in mammalian cells. Because Lin28B participates in the promotion and development of tumors mostly by blocking the let-7 tumor suppressor family members, we sought to explore the associated mechanisms to gain insights into how Lin28B might be decreased in human cancer cells to increase let-7 levels and reverse malignancy. We demonstrated that the histone acetyltransferase PCAF, via its cold shock domain, directly interacts with and subsequently acetylates Lin28B in lung adenocarcinoma-derived H1299 cells. RT-qPCR assays showed that both let-7a-1 and let-7g were increased in PCAF-transfected H1299 cells. Lin28B is acetylated by ectopic PCAF and translocates from the nucleus to the cytoplasm in H1299 cells. The effects of acetylated Lin28B on let-7a-1 and let-7g are similar to that of stable knockdown of Lin28B in H1299 cells. The new role of PCAF in mediating Lin28B acetylation and the specific release of its target microRNAs in H1299 cells may shed light on the potential application of let-7 in the clinical treatment of lung cancer patients.
Sigma-Aldrich
Nα-Acetyl-L-lysine
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)