Merck
In-Situ and Real-time Monitoring of Mechanochemical Preparation of Li
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-09-14)
Nikola Biliškov, Andreas Borgschulte, Krunoslav Užarević, Ivan Halasz, Stipe Lukin, Sanja Milošević, Igor Milanović, Jasmina Grbović Novaković
ABSTRACT
For the first time, in situ monitoring of uninterrupted mechanochemical synthesis of two bimetallic amidoboranes, M