Merck
  • Inhibitory effects of four neonicotinoid active ingredients on acetylcholine esterase activity.
Inhibitory effects of four neonicotinoid active ingredients on acetylcholine esterase activity.
Acta biologica Hungarica (2017-12-22)
János Győri, Anna Farkas, Oksana Stolyar, András Székács, Mária Mörtl, Ágnes Vehovszky
ABSTRACT
There is a great concern about the decline of pollinators, and neonicotinoids emerging bee disorders are assumed to play a significant role. Since changes in learning ability has been observed in honey bees exposed to some acetylcholine esterase (AChE) inhibitors, we therefore, tested in vitro the effect of four neonicotinoids on purified eel AChE. AChE activity was inhibited in a concentration-dependent manner, and calculated IC
Product Description
Supelco
Clothianidin, PESTANAL®, analytical standard
Supelco
Thiacloprid, PESTANAL®, analytical standard