- Nerve growth factor plays a role in the neurotherapeutic effect of a CD45
Nerve growth factor plays a role in the neurotherapeutic effect of a CD45
Cytotherapy (2017-12-25)
Galit Gincberg, Esther Shohami, Victoria Trembovler, Alexander G Alexandrovich, Philip Lazarovici, Uriel Elchalal
PMID29274773
ABSTRACT
Human umbilical cord blood (HUCB) is an important source of stem cells for therapy of hematopoietic disorders and is a potential therapy for various neurological disorders, including traumatic brain injury (TBI). The expression of nerve growth factor (NGF) and its receptors TrkA, p75 NGF and its receptors were detected on CD45 The HUCB CD45 These findings strengthen the role of NGF and its TrkA receptor in the HUCB CD45