Merck
  • IRF8-dependent molecular complexes control the Th9 transcriptional program.
IRF8-dependent molecular complexes control the Th9 transcriptional program.
Nature communications (2017-12-14)
Etienne Humblin, Marion Thibaudin, Fanny Chalmin, Valentin Derangère, Emeric Limagne, Corentin Richard, Richard A Flavell, Sandy Chevrier, Sylvain Ladoire, Hélène Berger, Romain Boidot, Lionel Apetoh, Frédérique Végran, François Ghiringhelli
ABSTRACT
Interferon regulatory factors (IRF) have critical functions in lymphoid development and in immune response regulation. Although many studies have described the function of IRF4 in CD4+ T cells, few have focused on the IRF4 homologue, IRF8. Here, we show that IRF8 is required for Th9 differentiation in vitro and in vivo. IRF8 functions through a transcription factor complex consisting of IRF8, IRF4, PU.1 and BATF, which binds to DNA and boosts Il9 transcription. By contrast, IRF8 deficiency promotes the expression of other genes such as Il4, as IRF8 dimerises with the transcriptional repressor ETV6 and inhibits Il4 expression. In vivo, IRF8 is essential for the anti-tumour effects of Th9 cells in mouse melanoma models. Our results show that IRF8 complexes boost the Th9 program and repress Il4 expression to modulate Th9 cell differentiation, thereby implicating IRF8 as a potential therapeutic target to affect Th9 responses in cancer therapy.
Sigma-Aldrich
SIS3, ≥98% (HPLC), powder
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green