- Applications of thermo-reversible pluronic F-127 gels in pharmaceutical formulations.
Applications of thermo-reversible pluronic F-127 gels in pharmaceutical formulations.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques (2007-01-09)
J J Escobar-Chávez, M López-Cervantes, A Naïk, Y N Kalia, D Quintanar-Guerrero, A Ganem-Quintanar
PMID17207417
ABSTRACT
It is, sometimes, desirable to maintain a constant plasma drug concentration within the therapeutically effective concentration range. The use of high viscosity hydromiscible vehicles such as hydrophilic gels, is one of various approaches for controlled drug delivery, and represents an important area of pharmaceutical research and development. Of these systems, Pluronic F-127 (PF-127) provides the pharmacist with an excellent drug delivery system for a number of routes of administration and is compatible with many different substances. Gels containing penetration enhancers have proven to be especially popular for administering anti-inflammatory medications since they are relatively easy to prepare and very efficacious.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) dimethacrylate, average Mn ~12,500
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate, average Mn ~12,500
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate, average Mn ~5,800