Merck
Mirror-image polymerase chain reaction.
Cell discovery (2017-10-21)
Wenjun Jiang, Baochang Zhang, Chuyao Fan, Min Wang, Jiaxing Wang, Qiang Deng, Xianyu Liu, Ji Chen, Jishen Zheng, Lei Liu, Ting F Zhu
ABSTRACT
The construction of mirror-image biological systems may open the next frontier for biomedical technology development and discovery. Here we have designed and chemically synthesized a mutant version of the thermostable
Sigma-Aldrich
Triisopropylsilane, 98%
Pricing and availability is not currently available.