Merck
  • Piperine Decreases Binding of Drugs to Human Plasma and Increases Uptake by Brain Microvascular Endothelial Cells.
Piperine Decreases Binding of Drugs to Human Plasma and Increases Uptake by Brain Microvascular Endothelial Cells.
Phytotherapy research : PTR (2017-09-28)
Raghvendra K Dubey, Brigitte Leeners, Bruno Imthurn, Gabriele Susanne Merki-Feld, Marinella Rosselli
ABSTRACT
We previously reported that piperine, an active alkaloidal principal of black and long peppers, enhances drug bioavailability by inhibiting drug metabolism. Another mechanism influencing drug availability/uptake is its free fraction. Since piperine is highly lipophilic, we hypothesize that it could also interact with drugs through binding displacement and influence their bioavailability. Accordingly, using equilibrium dialysis, we investigated whether piperine alters the binding of model drug ligands, that is flunitrazepam, diazepam, warfarin, salicylic acid, propranolol, lidocaine, and disopyramide to human plasma (n = 4). Since alterations in binding influence drug disposition, we also studied the effects of piperine on the uptake of plasma bound
Sigma-Aldrich
Phenazine, 98%