Merck
  • Subcutaneous white adipocytes express a light sensitive signaling pathway mediated via a melanopsin/TRPC channel axis.
Subcutaneous white adipocytes express a light sensitive signaling pathway mediated via a melanopsin/TRPC channel axis.
Scientific reports (2017-11-29)
Katarina Ondrusova, Mohammad Fatehi, Amy Barr, Zofia Czarnecka, Wentong Long, Kunimasa Suzuki, Scott Campbell, Koenraad Philippaert, Matthew Hubert, Edward Tredget, Peter Kwan, Nicolas Touret, Martin Wabitsch, Kevin Y Lee, Peter E Light
ABSTRACT
Subcutaneous white adipose tissue (scWAT) is the major fat depot in humans and is a central player in regulating whole body metabolism. Skin exposure to UV wavelengths from sunlight is required for Vitamin D synthesis and pigmentation, although it is plausible that longer visible wavelengths that penetrate the skin may regulate scWAT function. In this regard, we discovered a novel blue light-sensitive current in human scWAT that is mediated by melanopsin coupled to transient receptor potential canonical cation channels. This pathway is activated at physiological intensities of light that penetrate the skin on a sunny day. Daily exposure of differentiated adipocytes to blue light resulted in decreased lipid droplet size, increased basal lipolytic rate and alterations in adiponectin and leptin secretion. Our results suggest that scWAT function may be directly under the influence of ambient sunlight exposure and may have important implications for our current understanding of adipocyte biology. (150 words).
Sigma-Aldrich
Clemizole hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Glycerol Assay Kit, sufficient for 200 colorimetric or fluorometric tests
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
all trans-Retinal, powder, ≥98%
Pricing and availability is not currently available. Contact Technical Service