Merck
  • Repair of UV-Induced DNA Damage Independent of Nucleotide Excision Repair Is Masked by MUTYH.
Repair of UV-Induced DNA Damage Independent of Nucleotide Excision Repair Is Masked by MUTYH.
Molecular cell (2017-11-18)
Abdelghani Mazouzi, Federica Battistini, Sarah C Moser, Joana Ferreira da Silva, Marc Wiedner, Michel Owusu, Charles-Hugues Lardeau, Anna Ringler, Beatrix Weil, Jürgen Neesen, Modesto Orozco, Stefan Kubicek, Joanna I Loizou
ABSTRACT
DNA lesions caused by UV damage are thought to be repaired solely by the nucleotide excision repair (NER) pathway in human cells. Patients carrying mutations within genes functioning in this pathway display a range of pathologies, including an increased susceptibility to cancer, premature aging, and neurological defects. There are currently no curative therapies available. Here we performed a high-throughput chemical screen for agents that could alleviate the cellular sensitivity of NER-deficient cells to UV-induced DNA damage. This led to the identification of the clinically approved anti-diabetic drug acetohexamide, which promoted clearance of UV-induced DNA damage without the accumulation of chromosomal aberrations, hence promoting cellular survival. Acetohexamide exerted this protective function by antagonizing expression of the DNA glycosylase, MUTYH. Together, our data reveal the existence of an NER-independent mechanism to remove UV-induced DNA damage and prevent cell death.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gliquidone
Sigma-Aldrich
Anti-phospho-H2A.X (Ser139) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
N-acetylcysteine amide, ≥98% (HPLC)