Merck
  • Three new oleanane-type triterpenoid saponins from the seeds of Celosia cristata L.
Three new oleanane-type triterpenoid saponins from the seeds of Celosia cristata L.
Natural product research (2017-07-01)
Xin Liu, Jifa Zhang, Kai Guo, Airong Jia, Miansong Zhang, Yaping Shi, Changheng Liu, Linlin Xiao, Zhenliang Sun
ABSTRACT
Phytochemical investigation of the 1-butanol soluble fraction of 60% ethanol extract of the seeds of Celosia cristata L. led to the identification of three new oleanane-type triterpenoid saponins. Using
Sigma-Aldrich
L-(−)-Xylose, mixture of anomers, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Pricing and availability is not currently available.