Merck
  • A simple point of care test can indicate the need for periodontal therapy to reduce the risk for adverse pregnancy outcomes in mothers attending antenatal clinics.
A simple point of care test can indicate the need for periodontal therapy to reduce the risk for adverse pregnancy outcomes in mothers attending antenatal clinics.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2017-06-01)
Mervyn Sydney Turton, Ralf Reinhold Henkel, Charlene Wilma Joyce Africa
ABSTRACT
Although the association between periodontal disease (PD) and adverse pregnancy outcomes has gained recognition amongst antenatal healthcare workers, not much has changed in practice to address it. This prospective study tested the hypothesis that BANA (N-benzoyl-DL-arginine-2-naphthylamide), a diagnostic test for PD, may inform obstetricians and other antenatal healthcare practitioners, of the risk of adverse pregnancy outcomes in mothers attending antenatal clinics. At first visit, the presence of suspected periodontopathogens was assessed by BANA testing of dental plaque from 443 mothers attending antenatal clinics in KwaZulu-Natal, South Africa and an association later sought with pregnancy outcomes. The accuracy of BANA to predict adverse pregnancy outcomes was evaluated by the calculation of likelihood ratios. The study complied with the Declaration of Helsinki. Significant differences were found between pregnancy outcomes of BANA-negative and BANA-positive mothers (p < 0.0001). BANA showed sensitivity and negative predictive values of 87% and 91%; 75% and 78%; 87% and 94% in detecting low birth weight, preterm delivery, and preterm low birth weight delivery respectively. This study confirms that BANA may indicate the need for periodontal therapy to reduce the risk of adverse pregnancy outcomes and could form part of the routine antenatal examination.
Sigma-Aldrich
Nα-Benzoyl-DL-arginine β-naphthylamide hydrochloride, trypsin substrate
Pricing and availability is not currently available.