- A novel D
A novel D
American journal of physiology. Endocrinology and metabolism (2017-08-16)
M S Brook, D J Wilkinson, W K Mitchell, J L Lund, B E Phillips, N J Szewczyk, H Kainulainen, S Lensu, L G Koch, S L Britton, P L Greenhaff, K Smith, P J Atherton
PMID28811296