Merck
  • Oncogenic PIK3CA induces centrosome amplification and tolerance to genome doubling.
Oncogenic PIK3CA induces centrosome amplification and tolerance to genome doubling.
Nature communications (2017-11-25)
Inma M Berenjeno, Roberto Piñeiro, Sandra D Castillo, Wayne Pearce, Nicholas McGranahan, Sally M Dewhurst, Valerie Meniel, Nicolai J Birkbak, Evelyn Lau, Laurent Sansregret, Daniele Morelli, Nnennaya Kanu, Shankar Srinivas, Mariona Graupera, Victoria E R Parker, Karen G Montgomery, Larissa S Moniz, Cheryl L Scudamore, Wayne A Phillips, Robert K Semple, Alan Clarke, Charles Swanton, Bart Vanhaesebroeck
ABSTRACT
Mutations in PIK3CA are very frequent in cancer and lead to sustained PI3K pathway activation. The impact of acute expression of mutant PIK3CA during early stages of malignancy is unknown. Using a mouse model to activate the Pik3ca H1047R hotspot mutation in the heterozygous state from its endogenous locus, we here report that mutant Pik3ca induces centrosome amplification in cultured cells (through a pathway involving AKT, ROCK and CDK2/Cyclin E-nucleophosmin) and in mouse tissues, and increased in vitro cellular tolerance to spontaneous genome doubling. We also present evidence that the majority of PIK3CA H1047R mutations in the TCGA breast cancer cohort precede genome doubling. These previously unappreciated roles of PIK3CA mutation show that PI3K signalling can contribute to the generation of irreversible genomic changes in cancer. While this can limit the impact of PI3K-targeted therapies, these findings also open the opportunity for therapeutic approaches aimed at limiting tumour heterogeneity and evolution.
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Dihydrocytochalasin B
Sigma-Aldrich
Trypsin solution from porcine pancreas, 1 ×, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin per liter in Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse