- Membranous Nephropathy with an Enhanced Granular Expression of Thrombospondin Type-1 Domain-containing 7A in a Pregnant Woman.
Membranous Nephropathy with an Enhanced Granular Expression of Thrombospondin Type-1 Domain-containing 7A in a Pregnant Woman.
Internal medicine (Tokyo, Japan) (2016-09-16)
Takamasa Iwakura, Yoshihide Fujigaki, Naoko Katahashi, Taichi Sato, Sayaka Ishigaki, Naoko Tsuji, Yoshitaka Naito, Shinsuke Isobe, Masashi Ono, Yukitoshi Sakao, Takayuki Tsuji, Naro Ohashi, Akihiko Kato, Hiroaki Miyajima, Hideo Yasuda
PMID27629964
ABSTRACT
A 30-year-old woman with proteinuria first noted at 26 weeks of gestation was admitted to undergo further evaluation. A renal biopsy revealed membranous nephropathy (MN). There was no evidence of any secondary MN. Prednisolone was initiated 6 months after delivery. Four months later, her urine protein became negative. Enhanced granular staining for thrombospondin type-1 domain-containing 7A (THSD7A) in the glomeruli was retrospectively detected in a biopsy specimen. A literature review revealed that 60% of cases of THSD7A-related MN occurred in women of childbearing age. Therefore, THSD7A-related MN should be considered in female patients presenting with idiopathic MN in childbearing age.