- Imaging Monitoring of Kupffer Cell Function and Hepatic Oxygen Saturation in Preneoplastic Changes During Cholangiocarcinogenesis.
Imaging Monitoring of Kupffer Cell Function and Hepatic Oxygen Saturation in Preneoplastic Changes During Cholangiocarcinogenesis.
Scientific reports (2017-10-29)
Seunghyun Lee, Jung Hoon Kim, Jeong Hwa Lee, Yoh Zen, Joon Koo Han
PMID29079853
ABSTRACT
We investigated serial changes of the Kupffer cell (KC) function and hepatic oxygen saturation (sO
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution