- Quantitative Trait Loci and Maternal Effects Affecting the Strong Grain Dormancy of Wild Barley (
Quantitative Trait Loci and Maternal Effects Affecting the Strong Grain Dormancy of Wild Barley (
Frontiers in plant science (2017-11-23)
Shingo Nakamura, Mohammad Pourkheirandish, Hiromi Morishige, Mohammad Sameri, Kazuhiro Sato, Takao Komatsuda
PMID29163576
ABSTRACT
Wild barley (