Merck
  • NADPH oxidase-2 derived superoxide drives mitochondrial transfer from bone marrow stromal cells to leukemic blasts.
NADPH oxidase-2 derived superoxide drives mitochondrial transfer from bone marrow stromal cells to leukemic blasts.
Blood (2017-07-25)
Christopher R Marlein, Lyubov Zaitseva, Rachel E Piddock, Stephen D Robinson, Dylan R Edwards, Manar S Shafat, Zhigang Zhou, Matthew Lawes, Kristian M Bowles, Stuart A Rushworth
ABSTRACT
Improvements in the understanding of the metabolic cross-talk between cancer and its microenvironment are expected to lead to novel therapeutic approaches. Acute myeloid leukemia (AML) cells have increased mitochondria compared with nonmalignant CD34
Sigma-Aldrich
MISSION® esiRNA, targeting human CYBB