Merck
  • Evaluation of ecdysteroid antisera for a competitive enzyme immunoassay and extraction procedures for the measurement of mosquito ecdysteroids.
Evaluation of ecdysteroid antisera for a competitive enzyme immunoassay and extraction procedures for the measurement of mosquito ecdysteroids.
General and comparative endocrinology (2017-09-04)
David A McKinney, Michael R Strand, Mark R Brown
ABSTRACT
Ecdysteroid hormones regulate several aspects of insect development and reproduction. The predominant ecdysteroids produced by insects including mosquitoes are ecdysone (E) and 20-hydroxyecdysone (20E). The ability to measure E and 20E titers is essential for many studies, but few sensitive, low cost options are currently available for doing so. To address this deficiency, we developed a new enzyme-linked immunoassay (EIA). In the first part of the study, we compared the affinity of two new antisera named EAB25 and EAB27 to other available ecdysteroid antisera. EAB25 had a 27-fold higher affinity for 20E than E, while EAB27 had a four-fold higher affinity for 20E. In the second part of the study, EIA protocols were developed for analyzing E and 20E produced by the mosquito Aedes aegypti. Results indicated that pelts from fourth instar larvae and ovaries from blood-fed, adult females produced E and 20E. Methanol extraction in the presence of magnesium from whole body samples altered antibody recognition of E and 20E by EIA. However, extraction with 1-butanol and two organic/water phase separations eliminated this problem and improved assay performance. We conclude the new antisera used in the EIA provide a low-cost, flexible, and sensitive method for measuring E and 20E in insects.
Sigma-Aldrich
Ponasterone A, ≥65%
Sigma-Aldrich
7-Dehydrocholesterol, ≥95.0% (HPLC)