Merck
  • Hyperoxia causes miR-34a-mediated injury via angiopoietin-1 in neonatal lungs.
Hyperoxia causes miR-34a-mediated injury via angiopoietin-1 in neonatal lungs.
Nature communications (2017-10-29)
Mansoor Syed, Pragnya Das, Aishwarya Pawar, Zubair H Aghai, Anu Kaskinen, Zhen W Zhuang, Namasivayam Ambalavanan, Gloria Pryhuber, Sture Andersson, Vineet Bhandari
ABSTRACT
Hyperoxia-induced acute lung injury (HALI) is a key contributor to the pathogenesis of bronchopulmonary dysplasia (BPD) in neonates, for which no specific preventive or therapeutic agent is available. Here we show that lung micro-RNA (miR)-34a levels are significantly increased in lungs of neonatal mice exposed to hyperoxia. Deletion or inhibition of miR-34a improves the pulmonary phenotype and BPD-associated pulmonary arterial hypertension (PAH) in BPD mouse models, which, conversely, is worsened by miR-34a overexpression. Administration of angiopoietin-1, which is one of the downstream targets of miR34a, is able to ameliorate the BPD pulmonary and PAH phenotypes. Using three independent cohorts of human samples, we show that miR-34a expression is increased in type 2 alveolar epithelial cells in neonates with respiratory distress syndrome and BPD. Our data suggest that pharmacologic miR-34a inhibition may be a therapeutic option to prevent or ameliorate HALI/BPD in neonates.
Sigma-Aldrich
Anti-Angiopoietin-1 Antibody, from rabbit