Merck
  • The Scaffolding Protein IQGAP1 Interacts with NLRC3 and Inhibits Type I IFN Production.
The Scaffolding Protein IQGAP1 Interacts with NLRC3 and Inhibits Type I IFN Production.
Journal of immunology (Baltimore, Md. : 1950) (2017-09-03)
Aaron M Tocker, Emily Durocher, Kimberly D Jacob, Kate E Trieschman, Suzanna M Talento, Alma A Rechnitzer, David M Roberts, Beckley K Davis
ABSTRACT
Sensing of cytosolic nucleotides is a critical initial step in the elaboration of type I IFN. One of several upstream receptors, cyclic GMP-AMP synthase, binds to cytosolic DNA and generates dicyclic nucleotides that act as secondary messengers. These secondary messengers bind directly to stimulator of IFN genes (STING). STING recruits TNFR-associated NF-κB kinase-binding kinase 1 which acts as a critical node that allows for efficient activation of IFN regulatory factors to drive the antiviral transcriptome. NLRC3 is a recently characterized nucleotide-binding domain, leucine-rich repeat containing protein (NLR) that negatively regulates the type I IFN pathway by inhibiting subcellular redistribution and effective signaling of STING, thus blunting the transcription of type I IFNs. NLRC3 is predominantly expressed in lymphoid and myeloid cells. IQGAP1 was identified as a putative interacting partner of NLRC3 through yeast two-hybrid screening. In this article, we show that IQGAP1 associates with NLRC3 and can disrupt the NLRC3-STING interaction in the cytosol of human epithelial cells. Furthermore, knockdown of IQGAP1 in THP1 and HeLa cells causes significantly more IFN-β production in response to cytosolic nucleic acids. This result phenocopies NLRC3-deficient macrophages and fibroblasts and short hairpin RNA knockdown of NLRC3 in THP1 cells. Our findings suggest that IQGAP1 is a novel regulator of type I IFN production, possibly via interacting with NLRC3 in human monocytic and epithelial cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human IQGAP1