Merck
  • Zonula occludens-2 regulates Rho proteins activity and the development of epithelial cytoarchitecture and barrier function.
Zonula occludens-2 regulates Rho proteins activity and the development of epithelial cytoarchitecture and barrier function.
Biochimica et biophysica acta. Molecular cell research (2017-05-31)
Arturo Raya-Sandino, Alejandro Castillo-Kauil, Alaide Domínguez-Calderón, Lourdes Alarcón, David Flores-Benitez, Francisco Cuellar-Perez, Bruno López-Bayghen, Bibiana Chávez-Munguía, José Vázquez-Prado, Lorenza González-Mariscal
ABSTRACT
Silencing Zonula occludens 2 (ZO-2), a tight junctions (TJ) scaffold protein, in epithelial cells (MDCK ZO-2 KD) triggers: 1) Decreased cell to substratum attachment, accompanied by reduced expression of claudin-7 and integrin β1, and increased vinculin recruitment to focal adhesions and stress fibers formation; 2) Lowered cell-cell aggregation and appearance of wider intercellular spaces; 3) Increased RhoA/ROCK activity, mediated by GEF-HI recruitment to cell borders by cingulin; 4) Increased Cdc42 activity, mitotic spindle disorientation and the appearance of cysts with multiple lumens; 5) Increased Rac and cofilin activity, multiple lamellipodia formation and random cell migration but increased wound closure; 6) Diminished cingulin phosphorylation and disappearance of planar network of microtubules at the TJ region; and 7) Increased transepithelial electrical resistance at steady state, coupled to an increased expression of ZO-1 and claudin-4 and a decreased expression of claudin-2 and paracingulin. Hence, ZO-2 is a crucial regulator of Rho proteins activity and the development of epithelial cytoarchitecture and barrier function.
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
MISSION® esiRNA, targeting human AL358113.1, TJP2
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Anti-Integrin beta1 Antibody, Cytosolic, serum, Chemicon®
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection