- Continuous Production of Biorenewable, Polymer-Grade Lactone Monomers through Sn-β-Catalyzed Baeyer-Villiger Oxidation with H
Continuous Production of Biorenewable, Polymer-Grade Lactone Monomers through Sn-β-Catalyzed Baeyer-Villiger Oxidation with H
ChemSusChem (2017-08-15)
Keiko Yakabi, Thibault Mathieux, Kirstie Milne, Eva M López-Vidal, Antoine Buchard, Ceri Hammond
PMID28804968
ABSTRACT
The Baeyer-Villiger oxidation is a key transformation for sustainable chemical synthesis, especially when H