Merck
Palmitoylation of Ca
Biochemical and biophysical research communications (2017-07-26)
Abdulla S Kazim, Petter Storm, Enming Zhang, Erik Renström
ABSTRACT
High blood glucose triggers the release of insulin from pancreatic beta cells, but if chronic, causes cellular stress, partly due to impaired Ca
Sigma-Aldrich
2-Hydroxytetradecanoic acid, ≥98% (capillary GC)