Merck
  • MYC-Regulated Mevalonate Metabolism Maintains Brain Tumor-Initiating Cells.
MYC-Regulated Mevalonate Metabolism Maintains Brain Tumor-Initiating Cells.
Cancer research (2017-07-22)
Xiuxing Wang, Zhi Huang, Qiulian Wu, Briana C Prager, Stephen C Mack, Kailin Yang, Leo J Y Kim, Ryan C Gimple, Yu Shi, Sisi Lai, Qi Xie, Tyler E Miller, Christopher G Hubert, Anne Song, Zhen Dong, Wenchao Zhou, Xiaoguang Fang, Zhe Zhu, Vaidehi Mahadev, Shideng Bao, Jeremy N Rich
ABSTRACT
Metabolic dysregulation drives tumor initiation in a subset of glioblastomas harboring isocitrate dehydrogenase (IDH) mutations, but metabolic alterations in glioblastomas with wild-type IDH are poorly understood. MYC promotes metabolic reprogramming in cancer, but targeting MYC has proven notoriously challenging. Here, we link metabolic dysregulation in patient-derived brain tumor-initiating cells (BTIC) to a nexus between MYC and mevalonate signaling, which can be inhibited by statin or 6-fluoromevalonate treatment. BTICs preferentially express mevalonate pathway enzymes, which we find regulated by novel MYC-binding sites, validating an additional transcriptional activation role of MYC in cancer metabolism. Targeting mevalonate activity attenuated RAS-ERK-dependent BTIC growth and self-renewal. In turn, mevalonate created a positive feed-forward loop to activate MYC signaling via induction of miR-33b. Collectively, our results argue that MYC mediates its oncogenic effects in part by altering mevalonate metabolism in glioma cells, suggesting a therapeutic strategy in this setting. Cancer Res; 77(18); 4947-60. ©2017 AACR.
Sigma-Aldrich
6-Fluoromevalonate, ≥90% (GC), viscous liquid
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes