Merck
  • Absorption Kinetics of the Main Conjugated Linoleic Acid Isomers in Commercial-Rich Oil after Oral Administration in Rats.
Absorption Kinetics of the Main Conjugated Linoleic Acid Isomers in Commercial-Rich Oil after Oral Administration in Rats.
Journal of agricultural and food chemistry (2017-08-10)
Luís M Rodríguez-Alcalá, Irma Ares, Javier Fontecha, María-Rosa Martínez-Larrañaga, Arturo Anadón, María-Aránzazu Martínez
ABSTRACT
This study aimed to assess the oral absorption and plasma kinetics of two main isomers contained in commercial conjugated linoleic acid (CLA)-rich oil (Tonalin TG-80), rumenic acid (RA), and C18:2 trans-10, cis-12. The isomer plasma disposition after the single oral dose of 3000 mg of Tonalin TG-80/kg, containing 1200 mg/kg of each isomer, was studied in rats. The isomer plasma concentrations were determined by gas chromatography with flame ionization detection. The plasma kinetics showed rapid oral absorption of RA and C18:2 trans-10, cis-12 (t
Sigma-Aldrich
Glyceryl tritridecanoate, ≥99%