Merck
  • Stromal cell cadherin-11 regulates adipose tissue inflammation and diabetes.
Stromal cell cadherin-11 regulates adipose tissue inflammation and diabetes.
The Journal of clinical investigation (2017-08-02)
Sook Kyung Chang, Ayano C Kohlgruber, Fumitaka Mizoguchi, Xavier Michelet, Benjamin J Wolf, Kevin Wei, Pui Y Lee, Lydia Lynch, Danielle Duquette, Victòria Ceperuelo-Mallafré, Alexander S Banks, Michael B Brenner
ABSTRACT
M2 macrophages, innate lymphoid type 2 cells (ILC2s), eosinophils, Tregs, and invariant NK T cells (iNKT cells) all help to control adipose tissue inflammation, while M1 macrophages, TNF, and other inflammatory cytokines drive inflammation and insulin resistance in obesity. Stromal cells regulate leukocyte responses in lymph nodes, but the role of stromal cells in adipose tissue inflammation is unknown. PDGFRα+ stromal cells are major producers of IL-33 in adipose tissue. Here, we show that mesenchymal cadherin-11 modulates stromal fibroblast function. Cadherin-11-deficient mice displayed increased stromal production of IL-33, with concomitant enhancements in ILC2s and M2 macrophages that helped control adipose tissue inflammation. Higher expression levels of IL-33 in cadherin-11-deficient mice mediated ILC2 activation, resulting in higher IL-13 expression levels and M2 macrophage expansion in adipose tissue. Consistent with reduced adipose tissue inflammation, cadherin-11-deficient mice were protected from obesity-induced glucose intolerance and adipose tissue fibrosis. Importantly, anti-cadherin-11 mAb blockade similarly improved inflammation and glycemic control in obese WT mice. These results suggest that stromal fibroblasts expressing cadherin-11 regulate adipose tissue inflammation and thus highlight cadherin-11 as a potential therapeutic target for the management of obesity.
Sigma-Aldrich
Anti-OB-Cadherin Antibody, extracellular domain, clone 16A, clone 16A, Chemicon®, from mouse