Merck
  • Targeting Alpha-Fetoprotein (AFP)-MHC Complex with CAR T-Cell Therapy for Liver Cancer.
Targeting Alpha-Fetoprotein (AFP)-MHC Complex with CAR T-Cell Therapy for Liver Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2016-08-19)
Hong Liu, Yiyang Xu, Jingyi Xiang, Li Long, Shon Green, Zhiyuan Yang, Bryan Zimdahl, Jingwei Lu, Neal Cheng, Lucas H Horan, Bin Liu, Su Yan, Pei Wang, Juan Diaz, Lu Jin, Yoko Nakano, Javier F Morales, Pengbo Zhang, Lian-Xing Liu, Binnaz K Staley, Saul J Priceman, Christine E Brown, Stephen J Forman, Vivien W Chan, Cheng Liu
ABSTRACT
The majority of tumor-specific antigens are intracellular and/or secreted and therefore inaccessible by conventional chimeric antigen receptor (CAR) T-cell therapy. Given that all intracellular/secreted proteins are processed into peptides and presented by class I MHC on the surface of tumor cells, we used alpha-fetoprotein (AFP), a specific liver cancer marker, as an example to determine whether peptide-MHC complexes can be targets for CAR T-cell therapy against solid tumors. We generated a fully human chimeric antigen receptor, ET1402L1-CAR (AFP-CAR), with exquisite selectivity and specificity for the AFP We report that T cells expressing AFP-CAR selectively degranulated, released cytokines, and lysed liver cancer cells that were HLA-A*02:01 This study demonstrates that CAR T-cell immunotherapy targeting intracellular/secreted solid tumor antigens can elicit a potent antitumor response. Our approach expands the spectrum of antigens available for redirected T-cell therapy against solid malignancies and offers a promising new avenue for liver cancer immunotherapy. Clin Cancer Res; 23(2); 478-88. ©2016 AACR.
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)