Merck
  • Functional Cooperation between KCa3.1 and TRPV4 Channels in Bronchial Smooth Muscle Cell Proliferation Associated with Chronic Asthma.
Functional Cooperation between KCa3.1 and TRPV4 Channels in Bronchial Smooth Muscle Cell Proliferation Associated with Chronic Asthma.
Frontiers in pharmacology (2017-10-04)
Zhihua Yu, Yanxia Wang, Lu Qin, Hongzhuan Chen
ABSTRACT
Airway smooth muscle cells (SMC) proliferation contributes to the airways remodeling and irreversible airway obstruction during severe asthma, but the mechanisms of airway SMC proliferation are poorly understood. Intracellular Ca
Sigma-Aldrich
HC-067047, ≥98% (HPLC)