Merck
Coumarins from the roots of
Experimental and therapeutic medicine (2017-07-05)
Dong Li, Li Wu
ABSTRACT
Allergic inflammation is induced by allergens and leads to various allergic diseases, including rhinitis, asthma and conjunctivitis. Histamine is important in the pathogenesis of an immunoglobulin E-dependent allergic reaction and results in the secretion of cytokines associated with inflammation.
Sigma-Aldrich
Octadecylsilane, 97%