Merck
  • Metabolic Adaptation, a Specialized Leaf Organ Structure and Vascular Responses to Diurnal N
Metabolic Adaptation, a Specialized Leaf Organ Structure and Vascular Responses to Diurnal N
Frontiers in plant science (2017-04-15)
Paul Brouwer, Andrea Bräutigam, Valerie A Buijs, Anne O E Tazelaar, Adrie van der Werf, Urte Schlüter, Gert-Jan Reichart, Anthony Bolger, Björn Usadel, Andreas P M Weber, Henriette Schluepmann
ABSTRACT
Sustainable agriculture demands reduced input of man-made nitrogen (N) fertilizer, yet N
Sigma-Aldrich
Nitrogen-14N2, 99.99 atom % 14N