Merck
  • Resting-state Functional Connectivity is an Age-dependent Predictor of Motor Learning Abilities.
Resting-state Functional Connectivity is an Age-dependent Predictor of Motor Learning Abilities.
Cerebral cortex (New York, N.Y. : 1991) (2016-09-23)
Alison Mary, Vincent Wens, Marc Op de Beeck, Rachel Leproult, Xavier De Tiège, Philippe Peigneux
ABSTRACT
This magnetoencephalography study investigates how ageing modulates the relationship between pre-learning resting-state functional connectivity (rsFC) and subsequent learning. Neuromagnetic resting-state activity was recorded 5 min before motor sequence learning in 14 young (19-30 years) and 14 old (66-70 years) participants. We used a seed-based beta-band power envelope correlation approach to estimate rsFC maps, with the seed located in the right primary sensorimotor cortex. In each age group, the relation between individual rsFC and learning performance was investigated using Pearson's correlation analyses. Our results show that rsFC is predictive of subsequent motor sequence learning but involves different cross-network interactions in the two age groups. In young adults, decreased coupling between the sensorimotor network and the cortico-striato-cerebellar network is associated with better motor learning, whereas a similar relation is found in old adults between the sensorimotor, the dorsal-attentional and the DMNs. Additionally, age-related correlational differences were found in the dorsolateral prefrontal cortex, known to subtend attentional and controlled processes. These findings suggest that motor skill learning depends-in an age-dependent manner-on subtle interactions between resting-state networks subtending motor activity on the one hand, and controlled and attentional processes on the other hand.
Sigma-Aldrich
S-Methylisothiourea hemisulfate salt, 98%