Merck
  • Gender-Specific Amelioration of SMA Phenotype upon Disruption of a Deep Intronic Structure by an Oligonucleotide.
Gender-Specific Amelioration of SMA Phenotype upon Disruption of a Deep Intronic Structure by an Oligonucleotide.
Molecular therapy : the journal of the American Society of Gene Therapy (2017-04-17)
Matthew D Howell, Eric W Ottesen, Natalia N Singh, Rachel L Anderson, Ravindra N Singh
ABSTRACT
Spinal muscular atrophy (SMA), the leading genetic disease of children, is caused by low levels of survival motor neuron (SMN) protein. Here, we employ A15/283, an antisense oligonucleotide targeting a deep intronic sequence/structure, to examine the impact of restoration of SMN in a mild SMA mouse model. We show gender-specific amelioration of tail necrosis upon subcutaneous administrations of A15/283 into SMA mice at postnatal days 1 and 3. We also demonstrate that a modest increase in SMN due to early administrations of A15/283 dramatically improves testicular development and spermatogenesis. Our results reveal near total correction of expression of several genes in adult testis upon temporary increase in SMN during early postnatal development. This is the first demonstration of in vivo efficacy of an antisense oligonucleotide targeting a deep intronic sequence/structure. This is also the first report of gender-specific amelioration of SMA pathology upon a modest peripheral increase of SMN.
Sigma-Aldrich
Anti-SMN2 Antibody, clone SMN-KH, clone SMN-KH, from mouse