Merck
  • Colorimetry as Quality Control Tool for Individual Inkjet-Printed Pediatric Formulations.
Colorimetry as Quality Control Tool for Individual Inkjet-Printed Pediatric Formulations.
AAPS PharmSciTech (2016-10-16)
Henrika Wickström, Johan O Nyman, Mathias Indola, Heidi Sundelin, Leif Kronberg, Maren Preis, Jukka Rantanen, Niklas Sandler
ABSTRACT
Printing technologies were recently introduced to the pharmaceutical field for manufacturing of drug delivery systems. Printing allows on demand manufacturing of flexible pharmaceutical doses in a personalized manner, which is critical for a successful and safe treatment of patient populations with specific needs, such as children and the elderly, and patients facing multimorbidity. Printing of pharmaceuticals as technique generates new demands on the quality control procedures. For example, rapid quality control is needed as the printing can be done on demand and at the point of care. This study evaluated the potential use of a handheld colorimetry device for quality control of printed doses of vitamin Bs on edible rice and sugar substrates. The structural features of the substrates with and without ink were also compared. A multicomponent ink formulation with vitamin B
Sigma-Aldrich
Pyridoxine, ≥98%