Merck
Spiders have rich pigmentary and structural colour palettes.
The Journal of experimental biology (2017-06-02)
Bor-Kai Hsiung, Nicholas M Justyn, Todd A Blackledge, Matthew D Shawkey
ABSTRACT
Elucidating the mechanisms of colour production in organisms is important for understanding how selection acts upon a variety of behaviours. Spiders provide many spectacular examples of colours used in courtship, predation, defence and thermoregulation, but are thought to lack many types of pigments common in other animals. Ommochromes, bilins and eumelanin have been identified in spiders, but not carotenoids or melanosomes. Here, we combined optical microscopy, refractive index matching, confocal Raman microspectroscopy and electron microscopy to investigate the basis of several types of colourful patches in spiders. We obtained four major results. First, we show that spiders use carotenoids to produce yellow, suggesting that such colours may be used for condition-dependent courtship signalling. Second, we established the Raman signature spectrum for ommochromes, facilitating the identification of ommochromes in a variety of organisms in the future. Third, we describe a potential new pigmentary-structural colour interaction that is unusual because of the use of long wavelength structural colour in combination with a slightly shorter wavelength pigment in the production of red. Finally, we present the first evidence for the presence of melanosomes in arthropods, using both scanning and transmission electron microscopy, overturning the assumption that melanosomes are a synapomorphy of vertebrates. Our research shows that spiders have a much richer colour production palette than previously thought, and this has implications for colour diversification and function in spiders and other arthropods.
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
1,4-Naphthoquinone, 97%