Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Chronic pharmacological blockade of the Na
Chronic pharmacological blockade of the Na
The European journal of neuroscience (2017-07-18)
Pradeep Sherkhane, Josef P Kapfhammer
PMID
28715135
ABSTRACT
The Na
Product Description
K4144
Sigma-Aldrich
KB-R7943, ≥98% (HPLC), powder
Pricing
C127
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Pricing
B5016
Sigma-Aldrich
Bepridil hydrochloride, powder
Pricing