Merck
  • Ferroptosis and autophagy induced cell death occur independently after siramesine and lapatinib treatment in breast cancer cells.
Ferroptosis and autophagy induced cell death occur independently after siramesine and lapatinib treatment in breast cancer cells.
PloS one (2017-08-23)
Shumei Ma, Rebecca F Dielschneider, Elizabeth S Henson, Wenyan Xiao, Tricia R Choquette, Anna R Blankstein, Yongqiang Chen, Spencer B Gibson
ABSTRACT
Ferroptosis is a cell death pathway characterized by iron-dependent accumulation of reactive oxygen species (ROS) within the cell. The combination of siramesine, a lysosome disruptor, and lapatinib, a dual tyrosine kinase inhibitor, has been shown to synergistically induce cell death in breast cancer cells mediated by ferroptosis. These treatments also induce autophagy but its role in this synergistic cell death is unclear. In this study, we determined that siramesine and lapatinib initially induced ferroptosis but changes to an autophagy induced cell death after 24 hours. Furthermore, we found that intracellular iron level increased in a time dependent manner following treatment accompanied by an increase in ROS. Using the iron chelator deferoxamine (DFO) or the ROS scavenger alpha-tocopherol decreased both autophagy flux and cell death. We further discovered that decreased expression of the iron storage protein, ferritin was partially dependent upon autophagy degradation. In contrast, the expression of transferrin, which is responsible for the transport of iron into cells, is increased following treatment with lapatinib alone or in combination with siramesine. This indicates that ferroptosis and autophagy induced cell death occur independently but both are mediated by iron dependent ROS generation in breast cancer cells.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution