- Genome-Wide Identification and Functional Analysis of the Calcineurin B-like Protein and Calcineurin B-like Protein-Interacting Protein Kinase Gene Families in Turnip (
Genome-Wide Identification and Functional Analysis of the Calcineurin B-like Protein and Calcineurin B-like Protein-Interacting Protein Kinase Gene Families in Turnip (
Frontiers in plant science (2017-07-25)
Xin Yin, Qiuli Wang, Qian Chen, Nan Xiang, Yunqiang Yang, Yongping Yang
PMID28736570
ABSTRACT
The calcineurin B-like protein (CBL)-CBL-interacting protein kinase (CIPK) complex has been identified as a primary component in calcium sensors that perceives various stress signals. Turnip (