Merck
  • Crosstalk from cAMP to ERK1/2 emerges during postnatal maturation of nociceptive neurons and is maintained during aging.
Crosstalk from cAMP to ERK1/2 emerges during postnatal maturation of nociceptive neurons and is maintained during aging.
Journal of cell science (2017-05-19)
Joerg Isensee, Cosimo Schild, Frank Schwede, Tim Hucho
ABSTRACT
Maturation of nociceptive neurons depends on changes in transcription factors, ion channels and neuropeptides. Mature nociceptors initiate pain in part by drastically reducing the activation threshold via intracellular sensitization signaling. Whether sensitization signaling also changes during development and aging remains so far unknown. Using a novel automated microscopy approach, we quantified changes in intracellular signaling protein expression and in their signaling dynamics, as well as changes in intracellular signaling cascade wiring, in sensory neurons from newborn to senescent (24 months of age) rats. We found that nociceptive subgroups defined by the signaling components protein kinase A (PKA)-RIIβ (also known as PRKAR2B) and CaMKIIα (also known as CAMK2A) developed at around postnatal day 10, the time of nociceptor maturation. The integrative nociceptor marker, PKA-RIIβ, allowed subgroup segregation earlier than could be achieved by assessing the classical markers TRPV1 and Na
Sigma-Aldrich
4-Acetoxybenzyl alcohol, 97%