- Premature chromatid separation and altered proliferation of human leukocytes treated with vanadium (III) oxide.
Premature chromatid separation and altered proliferation of human leukocytes treated with vanadium (III) oxide.
Drug and chemical toxicology (2016-12-13)
Rodrigo Anibal Mateos-Nava, Juan José Rodríguez-Mercado, Mario Agustín Altamirano-Lozano
PMID27936973
ABSTRACT
Vanadium is a widely distributed metal in the Earth's surface and is released into the environment by either natural or anthropogenic causes. Vanadium (III) oxide (V
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)